Gene Transcripts

Gene Symbol
Evi5l
Entrez Gene ID
304201
Gene Name
ecotropic viral integration site 5-like
Gene Aliases
RGD1561962
Chromosome Location
Chr. 12: 2588066 - 2626318 on Build Rnor_6.0
UniGene ID
Rn.162082
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769048.1 XP_008767270.1 14 1668
GenBank mRNA FQ211110.1 - 12 1453

View Full Product Details