Gene Transcripts

Gene Symbol
Tssk5
Entrez Gene ID
315095
Gene Name
testis-specific serine kinase 5
Gene Aliases
RGD1308613
Chromosome Location
Chr. 7: 117437304 - 117440342 on Build Rnor_6.0
UniGene ID
Rn.222378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014086.2 NP_001014108.2 1 349
GenBank mRNA BC089206.1 - 1 329

View Full Product Details