Gene Transcripts

Gene Symbol
Prl5a1
Entrez Gene ID
171556
Gene Name
prolactin family 5, subfamily a, member 1
Gene Aliases
Prlpl
Chromosome Location
Chr. 17: 38323633 - 38331051 on Build Rnor_6.0
UniGene ID
Rn.18140
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138527.1 NP_612536.1 2 206
GenBank mRNA AB022883.1 - 2 206
AF226607.1 - 2 176
FQ220484.1 - 2 214
FQ230176.1 - 2 214

View Full Product Details