Gene Transcripts

Gene Symbol
Klra22
Entrez Gene ID
24933
Gene Name
killer cell lectin-like receptor subfamily A, member 22
Gene Aliases
Klra1, Ly49, Ly49s2
Chromosome Location
Chr. 4: 164384701 - 164406096 on Build Rnor_6.0
UniGene ID
Rn.10564
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173291.1 NP_775413.1 3 318
GenBank mRNA AY649838.1 - 3 329
U56822.1 - 3 318

View Full Product Details