Gene Transcripts

Gene Symbol
Iscu
Entrez Gene ID
288740
Gene Name
iron-sulfur cluster assembly enzyme
Gene Aliases
RGD1309562
Chromosome Location
Chr. 12: 48621454 - 48627297 on Build Rnor_6.0
UniGene ID
Rn.121
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105936.1 NP_001099406.1 1, 2 159
GenBank mRNA BC167056.1 - 1, 2 138

View Full Product Details