Gene Transcripts

Gene Symbol
Fam8a1
Entrez Gene ID
291031
Gene Name
family with sequence similarity 8, member A1
Gene Aliases
Rbm24
Chromosome Location
Chr. 17: 18413095 - 18421971 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771560.1 XP_008769782.1 4 1067
XM_214446.7 XP_214446.4 5 1201

View Full Product Details