Gene Transcripts

Gene Symbol
Wwox
Entrez Gene ID
292041
Gene Name
WW domain-containing oxidoreductase
Gene Aliases
Chromosome Location
Chr. 19: 46761368 - 47695247 on Build Rnor_6.0
UniGene ID
Rn.229354
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106188.1 NP_001099658.1 4 377
XM_006255666.2 XP_006255728.1 4 477
XM_006255670.2 XP_006255732.1 4 473
XM_006255671.2 XP_006255733.1 4 473
XM_006255673.2 XP_006255735.1 4 473
XM_008772491.1 XP_008770713.1 4 478

View Full Product Details