Gene Transcripts

Gene Symbol
Tubb5
Entrez Gene ID
29214
Gene Name
tubulin, beta 5 class I
Gene Aliases
Tubb
Chromosome Location
Chr. 20: 3422448 - 3426420 on Build Rnor_6.0
UniGene ID
Rn.2458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173102.2 NP_775125.1 4 949
GenBank mRNA AB011679.1 - 4 908
BC060540.1 - 3 394

View Full Product Details