Gene Transcripts

Gene Symbol
Fam103a1
Entrez Gene ID
293058
Gene Name
family with sequence similarity 103, member A1
Gene Aliases
RGD1310022
Chromosome Location
Chr. 1: 143583675 - 143589563 on Build Rnor_6.0
UniGene ID
Rn.40252
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127451.1 NP_001120923.1 2 174
GenBank mRNA BC089896.1 - 2 166
CF977463.1 - 2 194
DV717487.1 - 1 27

View Full Product Details