Gene Transcripts

Gene Symbol
Fhdc1
Entrez Gene ID
295161
Gene Name
FH2 domain containing 1
Gene Aliases
RGD1311955
Chromosome Location
Chr. 2: 183450608 - 183487820 on Build Rnor_6.0
UniGene ID
Rn.4218
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106437.1 NP_001099907.1 10 1404

View Full Product Details