Gene Transcripts

Gene Symbol
Plekhn1
Entrez Gene ID
298694
Gene Name
pleckstrin homology domain containing, family N member 1
Gene Aliases
RGD1311019
Chromosome Location
Chr. 5: 173646055 - 173654136 on Build Rnor_6.0
UniGene ID
Rn.207259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134523.1 NP_001127995.1 9 1379
XM_006239564.2 XP_006239626.1 9 1610
XM_006239565.2 XP_006239627.1 9 1610
XM_006239566.2 XP_006239628.1 9 1544

View Full Product Details