Gene Transcripts

Gene Symbol
Klhl14
Entrez Gene ID
364823
Gene Name
kelch-like family member 14
Gene Aliases
RGD1566178
Chromosome Location
Chr. 18: 12526627 - 12640381 on Build Rnor_6.0
UniGene ID
Rn.39343
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108885.1 NP_001102355.1 6 1344
XM_008771952.1 XP_008770174.1 3 1263

View Full Product Details