Gene Transcripts

Gene Symbol
Vom1r92
Entrez Gene ID
494310
Gene Name
vomeronasal 1 receptor 92
Gene Aliases
V1rb6
Chromosome Location
Chr. 4: 121758942 - 121761699 on Build Rnor_6.0
UniGene ID
Rn.129648
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008968.1 NP_001008968.1 1 203
XM_006236847.1 XP_006236909.1 2 278
GenBank mRNA AY510279.1 - 1 203

View Full Product Details