Gene Transcripts

Gene Symbol
Fbxo27
Entrez Gene ID
499114
Gene Name
F-box protein 27
Gene Aliases
RGD1563982
Chromosome Location
Chr. 1: 86872808 - 86878709 on Build Rnor_6.0
UniGene ID
Rn.105566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110491.1 NP_001103961.1 843
GenBank mRNA BC091204.1 - 843

View Full Product Details