Gene Transcripts

Gene Symbol
Rab19
Entrez Gene ID
500088
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 4: 67166412 - 67176141 on Build Rnor_6.0
UniGene ID
Rn.53184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024326.1 NP_001019497.1 2 152
XM_008762820.1 XP_008761042.1 2 580
GenBank mRNA BC088443.1 - 2 152

View Full Product Details