Gene Transcripts

Gene Symbol
Cyp2f4
Entrez Gene ID
54246
Gene Name
cytochrome P450, family 2, subfamily f, polypeptide 4
Gene Aliases
CYP4502F4, Cyp2f1, Cyp2f2
Chromosome Location
Chr. 1: 83933974 - 83947804 on Build Rnor_6.0
UniGene ID
Rn.10817
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019303.1 NP_062176.1 6 851
XM_006228585.2 XP_006228647.1 7 959
GenBank mRNA AF017393.1 - 6 851
BC070939.1 - 7 895 7099393

View Full Product Details