Gene Transcripts

Gene Symbol
N5
Entrez Gene ID
64825
Gene Name
DNA binding protein N5
Gene Aliases
Chromosome Location
Chr. 4: 57588724 - 57590923 on Build Rnor_6.0
UniGene ID
Rn.202953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022857.1 NP_074048.1 2 1167
GenBank mRNA L31882.1 - 2 1167

View Full Product Details