Gene Transcripts
- Gene Symbol
- Cdk3-ps
- Entrez Gene ID
- 69681
- Gene Name
- cyclin-dependent kinase 3, pseudogene
- Gene Aliases
- 2310050O10Rik, Cdk3
- Chromosome Location
- Chr. 11: 116216002 - 116220287 on Build GRCm38
- UniGene ID
- Mm.33677
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM934713.1 |
- |
2 |
66 |
|
Chr. 11 - 116216928 - 116219934 [+] |
GenBankmRNA |
BY709505.1 |
- |
3 |
350 |
|
Chr. 11 - 116216552 - 116219872 [+] |
View Full Product Details