Gene Transcripts
- Gene Symbol
- 2410018L13Rik
- Entrez Gene ID
- 69732
- Gene Name
- RIKEN cDNA 2410018L13 gene
- Gene Aliases
- 2810480C08Rik
- Chromosome Location
- Chr. 12: 22953997 - 23010243 on Build GRCm38
- UniGene ID
- Mm.391403
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015504.1 |
|
3 |
543 |
|
GenBank mRNA |
AK134640.1 |
- |
3 |
544 |
|
BC063063.1 |
- |
4 |
2513 |
|
- Gene Symbol
- Asap2
- Entrez Gene ID
- 211914
- Gene Name
- ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
- Gene Aliases
- 6530401G17Rik, Ddef2, Gm1523, Gm592, PAG3, PAP
- Chromosome Location
- Chr. 12: 21111713 - 21270171 on Build GRCm38
- UniGene ID
- Mm.358946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018309.1 |
- |
|
895 |
|
- Gene Symbol
- Gm9222
- Entrez Gene ID
- 668525
- Gene Name
- predicted gene 9222
- Gene Aliases
- BDR-1, CA748277, EG668383, EG668525
- Chromosome Location
- Chr. 12: 17881914 - 17939655 on Build GRCm38
- UniGene ID
- Mm.278091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031503.1 |
- |
|
1064 |
|
- Gene Symbol
- Hpcal1
- Entrez Gene ID
- 53602
- Gene Name
- hippocalcin-like 1
- Gene Aliases
- NVP-3, Nvp3, VILIP3, Vnsl3, Vsnl3
- Chromosome Location
- Chr. 12: 17690750 - 17791944 on Build GRCm38
- UniGene ID
- Mm.313619
- Species
- Mus musculus
View Full Product Details