Gene Transcripts
- Gene Symbol
- Cts8-ps
- Entrez Gene ID
- 116911
- Gene Name
- cathepsin 8, pseudogene
- Gene Aliases
- Cts2-ps
- Chromosome Location
- Chr. 13: 61281380 - 61288850 on Build GRCm38
- UniGene ID
- Mm.475565
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027871.2 |
|
3 |
1481 |
|
GenBank mRNA |
BC050022.1 |
- |
3 |
1481 |
|
View Full Product Details