Gene Transcripts
- Gene Symbol
- Ceacam-ps1
- Entrez Gene ID
- 100038912
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule pseudogene 1
- Gene Aliases
- Gm1978
- Chromosome Location
- Chr. 7: 16649030 - 16657478 on Build GRCm38
- UniGene ID
- Mm.389813
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003247.2 |
|
3 |
664 |
|
GenBank mRNA |
CK023612.1 |
- |
|
664 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK022275.1 |
- |
3 |
664 |
|
Chr. 7 - 16648934 - 16657478 [-] |
GenBankmRNA |
CK023298.1 |
- |
3 |
661 |
|
Chr. 7 - 16648954 - 16657476 [-] |
View Full Product Details