Gene Transcripts
- Gene Symbol
- Tubb2a-ps2
- Entrez Gene ID
- 627110
- Gene Name
- tubulin, beta 2a, pseudogene 2
- Gene Aliases
- C79445
- Chromosome Location
- Chr. 12: 11882196 - 11882899 on Build GRCm38
- UniGene ID
- Mm.194735
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003964.2 |
|
1 |
14 |
|
GenBank mRNA |
BG066514.2 |
- |
1 |
14 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
C79445.1 |
- |
|
11 |
|
Chr. - - [-] |
View Full Product Details