Gene Transcripts
- Gene Symbol
- Snord17
- Entrez Gene ID
- 100313519
- Gene Name
- small nucleolar RNA, C/D box 17
- Gene Aliases
- MBI-43
- Chromosome Location
- Chr. 2: 144265982 - 144266202 on Build GRCm38
- UniGene ID
- Mm.453538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030762.1 |
|
1 |
108 |
|
GenBank mRNA |
AF357317.1 |
- |
1 |
108 |
|
View Full Product Details