Gene Transcripts
- Gene Symbol
- Gorasp2
- Entrez Gene ID
- 70231
- Gene Name
- golgi reassembly stacking protein 2
- Gene Aliases
- 0610011A07Rik, 2410043M02Rik, 5730520M13Rik, 9430094F20Rik, AW552058, GOLPH2, GRASP55, GRS2, p59
- Chromosome Location
- Chr. 2: 70661489 - 70691725 on Build GRCm38
- UniGene ID
- Mm.271950
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY703029.1 |
- |
2 |
308 |
|
Chr. 2 - 70661625 - 70679559 [+] |
GenBankmRNA |
BI112264.1 |
- |
2 |
350 |
|
Chr. 2 - 70661587 - 70678616 [+] |
GenBankmRNA |
BY739978.1 |
- |
2 |
359 |
|
Chr. 2 - 70661576 - 70678087 [+] |
View Full Product Details