Gene Transcripts

Gene Symbol
Cyp2d13
Entrez Gene ID
68444
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 13
Gene Aliases
1300007K12Rik
Chromosome Location
Chr. 15: 82636750 - 82642045 on Build GRCm38
UniGene ID
Mm.443212
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003552.1 5 829
GenBank mRNA AK004933.1 - 659
BC018344.1 - 5 809


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI147110.1 - 2 251 Chr. 15 - 82636826 - 82640267 [-]

View Full Product Details