Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_002873.2 | 2 | 1257 | |||
NR_075094.1 | 2 | 1131 | ||||
GenBank mRNA | AK044313.1 | - | LIT3405 Vax2OS |
2 | 1131 | |
AY589787.1 | - | LIT3407 Vax2OS |
2 | 1261 |