Gene Transcripts
- Gene Symbol
- Rn5s
- Entrez Gene ID
- 19804
- Gene Name
- 5S RNA
- Gene Aliases
- Chromosome Location
- Chr. 8: 123598156 - 123598276 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030686.1 |
|
|
45 |
|
- Gene Symbol
- Gm22109
- Entrez Gene ID
- 100861494
- Gene Name
- predicted gene, 22109
- Gene Aliases
- Rn5s23
- Chromosome Location
- Chr. 8: 123606611 - 123606731 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046147.1 |
|
|
45 |
|
- Gene Symbol
- Gm22291
- Entrez Gene ID
- 100861501
- Gene Name
- predicted gene, 22291
- Gene Aliases
- Rn5s30
- Chromosome Location
- Chr. 8: 123618534 - 123618654 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046154.1 |
|
|
45 |
|
- Gene Symbol
- Gm23284
- Entrez Gene ID
- 100861502
- Gene Name
- predicted gene, 23284
- Gene Aliases
- Rn5s31
- Chromosome Location
- Chr. 8: 123620239 - 123620359 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046155.1 |
|
|
45 |
|
- Gene Symbol
- Gm25018
- Entrez Gene ID
- 100861480
- Gene Name
- predicted gene, 25018
- Gene Aliases
- Rn5s10
- Chromosome Location
- Chr. 8: 123565967 - 123566087 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046127.1 |
|
|
45 |
|
- Gene Symbol
- Gm25212
- Entrez Gene ID
- 100861499
- Gene Name
- predicted gene, 25212
- Gene Aliases
- Rn5s28
- Chromosome Location
- Chr. 8: 123615097 - 123615217 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046152.1 |
|
|
45 |
|
- Gene Symbol
- Gm26391
- Entrez Gene ID
- 100861488
- Gene Name
- predicted gene, 26391
- Gene Aliases
- Rn5s17
- Chromosome Location
- Chr. 8: 123593069 - 123593189 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046140.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s100
- Entrez Gene ID
- 100861471
- Gene Name
- nuclear encoded rRNA 5S 100
- Gene Aliases
- Rn5s2
- Chromosome Location
- Chr. 8: 123538784 - 123538904 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046118.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s103
- Entrez Gene ID
- 100861472
- Gene Name
- nuclear encoded rRNA 5S 103
- Gene Aliases
- Rn5s3
- Chromosome Location
- Chr. 8: 123543871 - 123543991 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046119.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s104
- Entrez Gene ID
- 100861473
- Gene Name
- nuclear encoded rRNA 5S 104
- Gene Aliases
- Rn5s4
- Chromosome Location
- Chr. 8: 123545576 - 123545696 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046120.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s105
- Entrez Gene ID
- 100861474
- Gene Name
- nuclear encoded rRNA 5S 105
- Gene Aliases
- Rn5s5
- Chromosome Location
- Chr. 8: 123547263 - 123547383 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046122.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s108
- Entrez Gene ID
- 100861475
- Gene Name
- nuclear encoded rRNA 5S 108
- Gene Aliases
- Rn5s6
- Chromosome Location
- Chr. 8: 123552362 - 123552482 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046123.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s110
- Entrez Gene ID
- 100861477
- Gene Name
- nuclear encoded rRNA 5S 110
- Gene Aliases
- Rn5s7
- Chromosome Location
- Chr. 8: 123555761 - 123555881 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046124.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s111
- Entrez Gene ID
- 100861478
- Gene Name
- nuclear encoded rRNA 5S 111
- Gene Aliases
- Rn5s8
- Chromosome Location
- Chr. 8: 123557488 - 123557608 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046125.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s113
- Entrez Gene ID
- 100861479
- Gene Name
- nuclear encoded rRNA 5S 113
- Gene Aliases
- Rn5s9
- Chromosome Location
- Chr. 8: 123560886 - 123561006 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046126.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s117
- Entrez Gene ID
- 100861481
- Gene Name
- nuclear encoded rRNA 5S 117
- Gene Aliases
- Rn5s11
- Chromosome Location
- Chr. 8: 123567635 - 123567755 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046128.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s121
- Entrez Gene ID
- 100861482
- Gene Name
- nuclear encoded rRNA 5S 121
- Gene Aliases
- Rn5s12
- Chromosome Location
- Chr. 8: 123574395 - 123574515 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046129.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s122
- Entrez Gene ID
- 100861483
- Gene Name
- nuclear encoded rRNA 5S 122
- Gene Aliases
- Rn5s13
- Chromosome Location
- Chr. 8: 123576074 - 123576194 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046130.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s123
- Entrez Gene ID
- 100861484
- Gene Name
- nuclear encoded rRNA 5S 123
- Gene Aliases
- Rn5s14
- Chromosome Location
- Chr. 8: 123577753 - 123577873 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046132.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s124
- Entrez Gene ID
- 100861486
- Gene Name
- nuclear encoded rRNA 5S 124
- Gene Aliases
- Rn5s15
- Chromosome Location
- Chr. 8: 123579448 - 123579568 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046133.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s128
- Entrez Gene ID
- 100861487
- Gene Name
- nuclear encoded rRNA 5S 128
- Gene Aliases
- Rn5s16
- Chromosome Location
- Chr. 8: 123586240 - 123586360 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046134.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s133
- Entrez Gene ID
- 100861489
- Gene Name
- nuclear encoded rRNA 5S 133
- Gene Aliases
- Rn5s18
- Chromosome Location
- Chr. 8: 123594752 - 123594872 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046141.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s134
- Entrez Gene ID
- 100861490
- Gene Name
- nuclear encoded rRNA 5S 134
- Gene Aliases
- Rn5s19
- Chromosome Location
- Chr. 8: 123596455 - 123596575 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046143.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s136
- Entrez Gene ID
- 100861491
- Gene Name
- nuclear encoded rRNA 5S 136
- Gene Aliases
- Rn5s20
- Chromosome Location
- Chr. 8: 123599859 - 123599979 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046144.1 |
|
1 |
45 |
|
- Gene Symbol
- n-R5s138
- Entrez Gene ID
- 100861492
- Gene Name
- nuclear encoded rRNA 5S 138
- Gene Aliases
- Rn5s21
- Chromosome Location
- Chr. 8: 123603245 - 123603365 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046145.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s139
- Entrez Gene ID
- 100861493
- Gene Name
- nuclear encoded rRNA 5S 139
- Gene Aliases
- Rn5s22
- Chromosome Location
- Chr. 8: 123604946 - 123605066 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046146.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s141
- Entrez Gene ID
- 100861495
- Gene Name
- nuclear encoded rRNA 5S 141
- Gene Aliases
- Rn5s24
- Chromosome Location
- Chr. 8: 123608318 - 123608438 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046148.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s142
- Entrez Gene ID
- 100861496
- Gene Name
- nuclear encoded rRNA 5S 142
- Gene Aliases
- Rn5s25
- Chromosome Location
- Chr. 8: 123610015 - 123610135 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046149.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s143
- Entrez Gene ID
- 100861497
- Gene Name
- nuclear encoded rRNA 5S 143
- Gene Aliases
- Rn5s26
- Chromosome Location
- Chr. 8: 123611713 - 123611833 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046150.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s144
- Entrez Gene ID
- 100861498
- Gene Name
- nuclear encoded rRNA 5S 144
- Gene Aliases
- Rn5s27
- Chromosome Location
- Chr. 8: 123613412 - 123613532 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046151.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s146
- Entrez Gene ID
- 100861500
- Gene Name
- nuclear encoded rRNA 5S 146
- Gene Aliases
- Rn5s29
- Chromosome Location
- Chr. 8: 123616808 - 123616928 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046153.1 |
|
|
45 |
|
- Gene Symbol
- n-R5s149
- Entrez Gene ID
- 100861503
- Gene Name
- nuclear encoded rRNA 5S 149
- Gene Aliases
- Rn5s32
- Chromosome Location
- Chr. 8: 123621926 - 123622046 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046156.1 |
|
1 |
45 |
|
View Full Product Details