Gene Transcripts
- Gene Symbol
- Aldoart2
- Entrez Gene ID
- 79459
- Gene Name
- aldolase 1 A, retrogene 2
- Gene Aliases
- 4933425L11Rik, Aldo1-ps1, Aldoa-ps1
- Chromosome Location
- Chr. 12: 55565205 - 55566896 on Build GRCm38
- UniGene ID
- Mm.392781
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006425.1 |
- |
1 |
460 |
|
AK016920.1 |
- |
1 |
440 |
|
EF662062.1 |
- |
1 |
387 |
|
View Full Product Details