Gene Transcripts
- Gene Symbol
- Rmst
- Entrez Gene ID
- 110333
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-coding RNA)
- Gene Aliases
- AI853140, C230053E11Rik, D930049J19Rik, Dmt2, M2, Ncrms
- Chromosome Location
- Chr. 10: 92081746 - 92165178 on Build GRCm38
- UniGene ID
- Mm.429764
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028262.1 |
|
13 |
2656 |
|
GenBank mRNA |
AK086758.1 |
- |
13 |
2650 |
|
AK136927.1 |
- |
9 |
3005 |
|
View Full Product Details