Gene Transcripts
- Gene Symbol
- Rmst
- Entrez Gene ID
- 110333
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-coding RNA)
- Gene Aliases
- AI853140, C230053E11Rik, D930049J19Rik, Dmt2, M2, Ncrms
- Chromosome Location
- Chr. 10: 92081746 - 92165178 on Build GRCm38
- UniGene ID
- Mm.429764
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028262.1 |
|
9, 10 |
954 |
|
GenBank mRNA |
AK086758.1 |
- |
9, 10 |
947 |
|
AK136927.1 |
- |
5, 6 |
1784 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ572455.1 |
- |
6, 7 |
610 |
|
Chr. 10 - 92083887 - 92164663 [-] |
View Full Product Details