Gene Transcripts
- Gene Symbol
- CYP4F30P
- Entrez Gene ID
- 100132708
- Gene Name
- cytochrome P450 family 4 subfamily F member 30, pseudogene
- Gene Aliases
- C2orf14
- Chromosome Location
- Chr. 2: 130680050 - 130685863 on Build GRCh38
- UniGene ID
- Hs.734862
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023391.1 |
|
5 |
2199 |
|
GenBank mRNA |
AL136789.1 |
- |
5 |
2199 |
|
- Gene Symbol
- CYP4F62P
- Entrez Gene ID
- 646802
- Gene Name
- cytochrome P450 family 4 subfamily F member 62, pseudogene
- Gene Aliases
- CYP4F43P
- Chromosome Location
- Chr. 2: 130435662 - 130441681 on Build GRCh38
- UniGene ID
- Hs.640308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103761.1 |
|
5 |
2372 |
|
View Full Product Details