Gene Transcripts

Gene Symbol
HCG9
Entrez Gene ID
10255
Gene Name
HLA complex group 9 (non-protein coding)
Gene Aliases
HCGIX, HCGIX4
Chromosome Location
Chr. 6: 29975115 - 29978403 on Build GRCh38
UniGene ID
Hs.656251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028032.1 1 326
GenBank mRNA AK289926.1 - 1 349
DA293452.1 - 1 349
X95289.1 - 1 326

View Full Product Details