Gene Transcripts

Gene Symbol
SEC61A2
Entrez Gene ID
55176
Gene Name
Sec61 translocon alpha 2 subunit
Gene Aliases
Chromosome Location
Chr. 10: 12129641 - 12169958 on Build GRCh38
UniGene ID
Hs.112955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024576.2 12 1480
NR_024577.2 13 1613
GenBank mRNA AK057532.1 - 13 1554
BC026179.1 - 12 1430 4823130

View Full Product Details