Gene Transcripts
- Gene Symbol
- DISC2
- Entrez Gene ID
- 27184
- Gene Name
- disrupted in schizophrenia 2 (non-protein coding)
- Gene Aliases
- DISC1-AS1, DISC1OS, NCRNA00015
- Chromosome Location
- Chr. 1: 231814626 - 231818517 on Build GRCh38
- UniGene ID
- Hs.720568
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002227.2 |
|
1 |
695 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF222981.3 |
- |
1 |
7122 |
|
Chr. 1 - 231809945 - 231824936 [-] |
View Full Product Details