Gene Transcripts

Gene Symbol
GGT3P
Entrez Gene ID
2679
Gene Name
gamma-glutamyltransferase 3 pseudogene
Gene Aliases
GGT3
Chromosome Location
Chr. 22: 18773689 - 18791961 on Build GRCh38
UniGene ID
Hs.595809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003267.1 10 1484
GenBank mRNA BC143607.1 - 6 1230

View Full Product Details