Gene Transcripts
- Gene Symbol
- POLR2J4
- Entrez Gene ID
- 84820
- Gene Name
- RNA polymerase II subunit J4, pseudogene
- Gene Aliases
- RPB11-phi
- Chromosome Location
- Chr. 7: 43940895 - 44019194 on Build GRCh38
- UniGene ID
- Hs.657028
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003655.3 |
|
16 |
3939 |
|
GenBank mRNA |
BX537878.1 |
- |
16 |
3894 |
|
View Full Product Details