Gene Transcripts
- Gene Symbol
- CELF5
- Entrez Gene ID
- 60680
- Gene Name
- CUGBP, Elav-like family member 5
- Gene Aliases
- BRUNOL-5, BRUNOL5, CELF-5
- Chromosome Location
- Chr. 19: 3224676 - 3297076 on Build GRCh38
- UniGene ID
- Hs.655747
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DC311931.1 |
- |
1 |
61 |
|
Chr. 19 - 3225485 - 3282142 [+] |
GenBankmRNA |
BX461674.2 |
- |
1 |
51 |
|
Chr. 19 - 3225495 - 3282502 [+] |
GenBankmRNA |
BP201493.1 |
- |
2 |
173 |
|
Chr. 19 - 3225282 - 3278103 [+] |
View Full Product Details