Gene Transcripts
- Gene Symbol
- SCARNA18
- Entrez Gene ID
- 677765
- Gene Name
- small Cajal body-specific RNA 18
- Gene Aliases
- SCARNA18A, U109
- Chromosome Location
- Chr. 5: 83064204 - 83064337 on Build GRCh38
- UniGene ID
- Hs.693283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003139.1 |
|
1 |
51 |
|
GenBank mRNA |
AM055742.1 |
- |
1 |
51 |
|
View Full Product Details