Gene Transcripts

Gene Symbol
MTMR9LP
Entrez Gene ID
339483
Gene Name
myotubularin related protein 9-like, pseudogene
Gene Aliases
MTMR9L
Chromosome Location
Chr. 1: 32231660 - 32241710 on Build GRCh38
UniGene ID
Hs.471067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026850.1 7 2241
GenBank mRNA AK055560.1 - 8 1973
AK096109.1 - 6 1793
AL833686.1 - 9 2541
BC016171.1 - 4 1086
BC041632.1 - 4 896


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB549336.1 - 2 374 Chr. 1 - 32231661 - 32232184 [-]
GenBankmRNA BM833152.1 - 4 511 Chr. 1 - 32231698 - 32234981 [-]

View Full Product Details