Gene Transcripts
- Gene Symbol
- CFL1P1
- Entrez Gene ID
- 142913
- Gene Name
- cofilin 1 pseudogene 1
- Gene Aliases
- CFLP1
- Chromosome Location
- Chr. 10: 87818313 - 87845612 on Build GRCh38
- UniGene ID
- Hs.369431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028492.1 |
|
4 |
1267 |
|
GenBank mRNA |
BC031631.1 |
- |
4 |
1267 |
|
View Full Product Details