Gene Transcripts
- Gene Symbol
- FMO6P
- Entrez Gene ID
- 388714
- Gene Name
- flavin containing monooxygenase 6 pseudogene
- Gene Aliases
- FMO6
- Chromosome Location
- Chr. 1: 171137740 - 171161563 on Build GRCh38
- UniGene ID
- Hs.448988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002601.1 |
|
1 |
657 |
|
GenBank mRNA |
AK130511.1 |
- |
1 |
657 |
|
View Full Product Details