Gene Transcripts

Gene Symbol
FMO6P
Entrez Gene ID
388714
Gene Name
flavin containing monooxygenase 6 pseudogene
Gene Aliases
FMO6
Chromosome Location
Chr. 1: 171137740 - 171161563 on Build GRCh38
UniGene ID
Hs.448988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002601.1 1 746
GenBank mRNA AK130511.1 - 1 746

View Full Product Details