Gene Transcripts
- Gene Symbol
- KRT8P41
- Entrez Gene ID
- 283102
- Gene Name
- keratin 8 pseudogene 41
- Gene Aliases
- Chromosome Location
- Chr. 11: 9094363 - 9096190 on Build GRCh38
- UniGene ID
- Hs.689472
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027713.1 |
|
1 |
551 |
|
GenBank mRNA |
AK127993.1 |
- |
1 |
551 |
|
View Full Product Details