Gene Transcripts

Gene Symbol
MDS2
Entrez Gene ID
259283
Gene Name
myelodysplastic syndrome 2 translocation associated
Gene Aliases
Chromosome Location
Chr. 1: 23627334 - 23640566 on Build GRCh38
UniGene ID
Hs.523369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027042.1 4 564
GenBank mRNA AJ310434.2 - 4 564
BC041472.1 - 5 1459
BC069164.1 - 4 564

View Full Product Details