Gene Transcripts
- Gene Symbol
- OR3A4P
- Entrez Gene ID
- 390756
- Gene Name
- olfactory receptor family 3 subfamily A member 4 pseudogene
- Gene Aliases
- OLFRA05, OLFRA06, OR17-13, OR17-16, OR17-24, OR17-25, OR24, OR25, OR3A4, OR3A5P
- Chromosome Location
- Chr. 17: 3310245 - 3311446 on Build GRCh38
- UniGene ID
- Hs.632245
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024128.1 |
|
1 |
543 |
|
GenBank mRNA |
BC136874.1 |
- |
1 |
543 |
|
BC136875.1 |
- |
1 |
543 |
|
View Full Product Details