Gene Transcripts
- Gene Symbol
- HERC2P4
- Entrez Gene ID
- 100289574
- Gene Name
- hect domain and RLD 2 pseudogene 4
- Gene Aliases
- D16F37S5
- Chromosome Location
- Chr. 16: 32170043 - 32188113 on Build GRCh38
- UniGene ID
- Hs.531536
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038215.2 |
- |
2 |
598 |
|
View Full Product Details