Gene Transcripts

Gene Symbol
TMCC1
Entrez Gene ID
23023
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
Chromosome Location
Chr. 3: 129647792 - 129893605 on Build GRCh38
UniGene ID
Hs.477547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033361.1 1 110
XM_006713552.3 XP_006713615.1 1 155
GenBank mRNA AK124671.1 - 1 111
BC030619.2 - 1 28
BC047517.1 - 1 28
DB085398.1 - 1 35

View Full Product Details