Gene Transcripts

Gene Symbol
NXF5
Entrez Gene ID
55998
Gene Name
nuclear RNA export factor 5
Gene Aliases
Chromosome Location
Chr. X: 101832112 - 101857577 on Build GRCh38
UniGene ID
Hs.307077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028089.1 17 1706
GenBank mRNA AJ277654.1 - 17 1706
AJ277655.1 - 17 1681
AJ277656.1 - 17 1617
AJ277657.1 - 15 1492
AJ277658.1 - 15 1611

View Full Product Details