Gene Transcripts
- Gene Symbol
- TTTY19
- Entrez Gene ID
- 252952
- Gene Name
- testis-specific transcript, Y-linked 19 (non-protein coding)
- Gene Aliases
- NCRNA00144
- Chromosome Location
- Chr. Y: 8704472 - 8705283 on Build GRCh38
- UniGene ID
- Hs.522853
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001549.1 |
|
2 |
169 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF527831.3 |
- |
2 |
169 |
|
Chr. Y - 8704472 - 8705283 [+] |
View Full Product Details