Gene Transcripts
- Gene Symbol
- FTLP10
- Entrez Gene ID
- 100130017
- Gene Name
- ferritin light chain pseudogene 10
- Gene Aliases
- Chromosome Location
- Chr. 4: 68182292 - 68212470 on Build GRCh38
- UniGene ID
- Hs.582037
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015446.1 |
|
1 |
1074 |
|
GenBank mRNA |
AY885668.1 |
- |
1 |
1074 |
|
View Full Product Details